accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6421-04-1

IUPAC Name: 1,2-bis[4-(ethoxycarbonyl)phenyl]-2-oxohydrazin-2-ium-1-ide
Synonyms: 1,2-bis[4-(ethoxycarbonyl)phenyl]-2-oxohydrazin-2-ium-1-ide
Molecular Weight (g/mol): 342.35
Molecular Formula: C18H18N2O5
InChi Key: LOOVRYZFUGHEMF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C([N-][N+](=O)C2=CC=C(C=C2)C(=O)OCC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results