Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Azoxybenzene-4,4'-dicarboxylate 97.0+%, TCI America™
SDP

Catalog No. A10651G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10651G Supplier TCI America Supplier No. A10651G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6421-04-1
Molecular Formula C18H18N2O5
Molecular Weight (g/mol) 342.35
MDL Number MFCD00053758
InChI Key LOOVRYZFUGHEMF-UHFFFAOYSA-N
Synonym Azoxybenzene-4,4′C-dicarboxylic Acid Diethyl Ester
PubChem CID 80889
IUPAC Name (4-ethoxycarbonylphenyl)-(4-ethoxycarbonylphenyl)imino-oxidoazanium
SMILES CCOC(=O)C1=CC=C(C=C1)N=[N+](C2=CC=C(C=C2)C(=O)OCC)[O-]

Specifications

Melting Point 115°C
Color Yellow
Quantity 1 g
Formula Weight 342.35
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl Azoxybenzene-4,4′-dicarboxylate
EINECSNumber (3)-3025
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.