accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOONOO

CAS RN 6422-83-9

IUPAC Name: 1-[5-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-2-methylphenyl]-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-[5-(2,5-dioxopyrrol-1-yl)-2-methylphenyl]pyrrole-2,5-dione
Molecular Weight (g/mol): 282.26
Molecular Formula: C15H10N2O4
InChi Key: FJKKJQRXSPFNPM-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1N1C(=O)C=CC1=O)N1C(=O)C=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results