Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene 2,4-Dimaleimide, MP Biomedicals

Catalog No. ICN225691 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN225691 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN225691 Supplier MP Biomedicals, Inc Supplier No. 0522569105

Chemical Identifiers

CAS 6422-83-9
Molecular Formula C15H10N2O4
Molecular Weight (g/mol) 282.255
InChI Key FJKKJQRXSPFNPM-UHFFFAOYSA-N
Synonym 2,4-bismaleimidotoluene, 2,4-dimaleimidotoluene, toluene-2,4-bismaleimide, bismaleimide, 2,4-toluene, 2,4-tolylenebis maleimide, n,n'-2,4-toluenebismaleimide, toluene-2,4-dimaleimide, 1,3-bismaleimide-4-methylbenzene, n,n'-4-methyl-1,3-phenylene bismaleimide, 1,1'-4-methyl-1,3-phenylene bis-1h-pyrrole-2,5-dione
PubChem CID 80893
IUPAC Name 1-[3-(2,5-dioxopyrrol-1-yl)-4-methylphenyl]pyrrole-2,5-dione
SMILES CC1=C(C=C(C=C1)N2C(=O)C=CC2=O)N3C(=O)C=CC3=O

Specifications

Quantity 10 g
Chemical Name or Material Toluene-2, 4-Dimaleimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.