accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SSSNN

CAS RN 64224-21-1

IUPAC Name: 4-methyl-5-(pyrazin-2-yl)-3H-1,2-dithiole-3-thione
Synonyms: oltipraz 奥替普拉
Molecular Weight (g/mol): 226.33
Molecular Formula: C8H6N2S3
InChi Key: CKNAQFVBEHDJQV-UHFFFAOYSA-N
SMILES: CC1=C(SSC1=S)C1=CN=CC=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results