accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6425-39-4

IUPAC Name: 4-{2-[2-(morpholin-4-yl)ethoxy]ethyl}morpholine
Synonyms: 4-{2-[2-(morpholin-4-yl)ethoxy]ethyl}morpholine
Molecular Weight (g/mol): 244.34
Molecular Formula: C12H24N2O3
InChi Key: ZMSQJSMSLXVTKN-UHFFFAOYSA-N
SMILES: C(CN1CCOCC1)OCCN1CCOCC1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)