Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,4'-(Oxydi-2,1-ethanediyl)bismorpholine

Catalog No. 424528100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-452-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-452-8100ML Supplier Sigma Aldrich Supplier No. 424528100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,4G-(Oxydi-2,1-ethanediyl)bismorpholine (DMDEE) is an amine based catalyst that is also known as dimorpholino-diethyl ether. It can act as a catalyst for blowing reactions and facilitates the process of polymeric curing.DMDEE can be used as a property modifier for 3-nitribenzonitrile (3-NDN) which can be further used in matrix assisted ionization vacuum analysis (MAIV). It can also be used as catalyst for formation of polyurethane foams, adhesives and polypropylene glycol (PPG) incorporated fumed silica.

Specifications

CAS 6425-39-4
Density 1.06 g/mL (at 25°C)
Boiling Point 309°C
Molecular Formula C12H24N2O3
Refractive Index n20/D 1.484 (literature)
Linear Formula C12H24N2O3
MDL Number MFCD00072740
Quantity 100 mL
Synonym beta,beta-Dimorpholinodiethyl ether; 2,2 -Dimorpholinodiethyl ether; 4,4 -(Oxydiethylene)dimorpholine; Bis(morpholinoethyl) ether
Molecular Weight (g/mol) 244.33
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.