accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3COONOCH3H3C

CAS RN 64257-84-7

IUPAC Name: cyano(3-phenoxyphenyl)methyl 2,2,3,3-tetramethylcyclopropane-1-carboxylate
Synonyms: Rody (+-)-fenpropathrin herald fenpropathrin kilumal danitol epa pesticide chemical code 127901 danimen fenpropanate meiothrin
Molecular Weight (g/mol): 349.43
Molecular Formula: C22H23NO3
InChi Key: XQUXKZZNEFRCAW-UHFFFAOYNA-N
SMILES: CC1(C)C(C(=O)OC(C#N)C2=CC=CC(OC3=CC=CC=C3)=C2)C1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results