accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6427-66-3

IUPAC Name: 4-azidobenzoic acid
Synonyms: 4-azidobenzoic acid
Molecular Weight (g/mol): 163.14
Molecular Formula: C7H5N3O2
InChi Key: PQXPAFTXDVNANI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)N=[N+]=[N-]
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)