Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-{2-[2-(2-Mercaptoethoxy)Ethoxy]Ethoxy}Ethanol

Catalog No. 767751500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-775-1500MG 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 76-775-1500MG Supplier Sigma Aldrich Supplier No. 767751500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-{2-[2-(2-Mercaptoethoxy)ethoxy]ethoxy}ethanol may be used in the synthesis of 2-(2-(2-((7-nitrobenzo[c][1,2,5]oxadiazol-4-yl)thio)ethoxy)ethoxy)ethanol, an nitrobenzoxadiazole (NBD) analog of the anticancer agent 6-((7-nitrobenzo[c][1,2,5]oxadiazol-4-yl)thio)hexan-1-ol (NBDHEX). This analog is a potent glutathione S-transferase (GST) and a potential therapeutic antimelanoma agent with enhanced water solubility and antitumor efficacy when compared to NBDHEX in a study.

Specifications

CAS 6427-66-3
Molecular Formula C7H5N3O2
Linear Formula C7H5N3O2
MDL Number MFCD00013987
Quantity 10 mL
Synonym MEEE; 1-Mercapto-11-hydroxy-3,6,9-trioxaundecane
Molecular Weight (g/mol) 163.13 g/mol
Biological Activity Blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.