accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64287-19-0

IUPAC Name: 1-(4-fluoro-3-methoxyphenyl)ethan-1-one
Synonyms: 1-(4-fluoro-3-methoxyphenyl)ethanone
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: PFEGFUCYOHBDJF-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(=C1)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results