accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHOHOHOHHO

CAS RN 643-03-8

IUPAC Name: hexane-1,2,3,4,5,6-hexol
Synonyms: mannitol sorbo sorbitol D(-)-mannitol 甘露醇
Molecular Weight (g/mol): 182.17
Molecular Formula: C6H14O6
InChi Key: FBPFZTCFMRRESA-UHFFFAOYNA-N
SMILES: OCC(O)C(O)C(O)C(O)CO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results