accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOO

CAS RN 643-75-4

IUPAC Name: diphenylpropane-1,2,3-trione
Synonyms: diphenylpropane-1,2,3-trione
Molecular Weight (g/mol): 238.24
Molecular Formula: C15H10O3
InChi Key: RXVBJUZEFSAYPW-UHFFFAOYSA-N
SMILES: O=C(C(=O)C1=CC=CC=C1)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results