Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diphenylpropanetrione 98.0+%, TCI America™
SDP

Catalog No. D23255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23255G 5 g
D23251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23255G Supplier TCI America Supplier No. D23255G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 643-75-4
Molecular Formula C15H10O3
Molecular Weight (g/mol) 238.24
MDL Number MFCD00191426
InChI Key RXVBJUZEFSAYPW-UHFFFAOYSA-N
Synonym Dibenzoyl Ketone
PubChem CID 223850
IUPAC Name diphenylpropane-1,2,3-trione
SMILES O=C(C(=O)C1=CC=CC=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 71°C
Color Yellow
Quantity 5 g
Formula Weight 238.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diphenylpropanetrione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.