accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OHNNOHNNNOHOHOHOH2O

CAS RN 64350-24-9

IUPAC Name: N-{9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6-oxo-6,9-dihydro-3H-purin-2-yl}-2-methylpropanamide hydrate
Synonyms: N-{9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6-oxo-3H-purin-2-yl}-2-methylpropanamide hydrate
Molecular Weight (g/mol): 371.35
Molecular Formula: C14H21N5O7
InChi Key: JXIMAIWXKDBJKP-UHFFFAOYNA-N
SMILES: O.CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C1OC(CO)C(O)C1O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)