accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 644-49-5

H3COOCH3CH3

CAS RN 644-49-5

IUPAC Name: propyl 2-methylpropanoate
Synonyms: propyl isobutyrate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: AZFUASHXSOTBNU-UHFFFAOYSA-N
SMILES: CCCOC(=O)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results