Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl Isobutyrate 97.0+%, TCI America™
SDP

Catalog No. I010825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I010825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I010825ML Supplier TCI America Supplier No. I010825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 644-49-5
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00053781
InChI Key AZFUASHXSOTBNU-UHFFFAOYSA-N
Synonym Isobutyric Acid Propyl Ester
PubChem CID 12571
IUPAC Name propyl 2-methylpropanoate
SMILES CCCOC(=O)C(C)C

Specifications

Color Colorless
Boiling Point 134°C
Quantity 25 mL
UN Number 3272
Formula Weight 130.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Isobutyrate
EINECSNumber (2)-0779
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.