accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 6443-85-2

IUPAC Name: 2-(pyridin-3-yl)acetonitrile
Synonyms: 2-(pyridin-3-yl)acetonitrile
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: OIPHWUPMXHQWLR-UHFFFAOYSA-N
SMILES: N#CCC1=CC=CN=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (4)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)