Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyridylacetonitrile, MP Biomedicals

Catalog No. ICN218234 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218234 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218234 Supplier MP Biomedicals, Inc Supplier No. 0521823405

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6443-85-2
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00006406
InChI Key OIPHWUPMXHQWLR-UHFFFAOYSA-N
Synonym 3-pyridineacetonitrile, 3-pyridylacetonitrile, 2-pyridin-3-yl acetonitrile, pyridine-3-acetonitrile, 3-cyanomethylpyridine, 3-cyanomethyl pyridine, pyridin-3-ylacetonitrile, 2-3-pyridyl ethanenitrile, pyridin-3-yl-acetonitrile, 3-pyridyl acetonitrile
PubChem CID 80923
IUPAC Name 2-(pyridin-3-yl)acetonitrile
SMILES N#CCC1=CC=CN=C1

Specifications

Quantity 10 g
Formula Weight 118.14
Chemical Name or Material 3-Pyridylacetonitrile
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.