CAS RN 64439-81-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Camptothecin, 10-Hydroxy-, Camptotheca acuminata
A cell-permeable powerful DNA topoisomerase I inhibitor.