Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Camptothecin, 10-Hydroxy-, Camptotheca acuminata

Catalog No. 39023825MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-023-825MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-023-825MG Supplier MilliporeSigma™ Supplier No. 39023825MG
Add to Cart
Add to Cart

A cell-permeable powerful DNA topoisomerase I inhibitor.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 95% by HPLC
Target Proteins DNA topoisomerase I
CAS 64439-81-2
Molecular Formula C20H16N2O5
Formula Weight 364.4g/mol
Solubility DMSO
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.