accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNOONHNOO(Z)

CAS RN 6448-95-9

IUPAC Name: (4Z)-4-[2-(2-methyl-5-nitrophenyl)hydrazin-1-ylidene]-3-oxo-N-phenyl-3,4-dihydronaphthalene-2-carboxamide
Synonyms: (4Z)-4-[2-(2-methyl-5-nitrophenyl)hydrazin-1-ylidene]-3-oxo-N-phenylnaphthalene-2-carboxamide
Molecular Weight (g/mol): 426.43
Molecular Formula: C24H18N4O4
InChi Key: HVBDWKMAUMVFIY-QYQHSDTDSA-N
SMILES: CC1=CC=C(C=C1N\N=C1/C(=O)C(=CC2=CC=CC=C12)C(=O)NC1=CC=CC=C1)[N+]([O-])=O
Color 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)