accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64481-10-3

ClClOOO

CAS RN 64481-10-3

IUPAC Name: 6,8-dichloro-4-oxo-4H-chromene-3-carbaldehyde
Synonyms: 6,8-dichloro-4-oxochromene-3-carbaldehyde
Molecular Weight (g/mol): 243.04
Molecular Formula: C10H4Cl2O3
InChi Key: IHCCHRKNCOFDAJ-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C2OC=C(C=O)C(=O)C2=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)