Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,8-Dichlorochromone-3-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. D43885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43885G 5 g
D43881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43885G Supplier TCI America Supplier No. D43885G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64481-10-3
Molecular Formula C10H4Cl2O3
Molecular Weight (g/mol) 243.04
MDL Number MFCD00051510
InChI Key IHCCHRKNCOFDAJ-UHFFFAOYSA-N
Synonym 6,8-dichloro-3-formylchromone, 6,8-dichloro-4-oxo-4h-chromene-3-carbaldehyde, 6,8-dichlorochromone-3-carboxaldehyde, chembl85493, 6,8-dichloro-4-oxo-4h-1-benzopyran-3-carboxaldehyde, 4h-1-benzopyran-3-carboxaldehyde, 6,8-dichloro-4-oxo, pubchem8676, 6,8-dichlorochromon-3-aldehyde, 6,8-dichloro-4-oxo-chromene-3-carbaldehyde, 3-formyl-6,8-dichloro-4h-1-benzopyran-4-one
PubChem CID 522129
IUPAC Name 6,8-dichloro-4-oxo-4H-chromene-3-carbaldehyde
SMILES ClC1=CC(Cl)=C2OC=C(C=O)C(=O)C2=C1

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 5 g
Formula Weight 243.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6,8-Dichlorochromone-3-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.