accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3O(E)

CAS RN 645-62-5

IUPAC Name: (2E)-2-ethylhex-2-enal
Synonyms: 2-ethyl-2-hexenal
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: PYLMCYQHBRSDND-SOFGYWHQSA-N
SMILES: CCC\C=C(/CC)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results