Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-2-hexenal (cis- and trans- mixture), TCI America™
SDP

Catalog No. E0123500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0123500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E0123500ML Supplier TCI America Supplier No. E0123500ML

missing translation for 'validMainframeMsg'

Specifications

CAS 645-62-5
Color Yellow
Boiling Point 175°C
Molecular Formula C8H14O
MDL Number MFCD00006983
Quantity 500 mL
InChI Key PYLMCYQHBRSDND-VURMDHGXSA-N
SMILES CCCC=C(CC)C=O
IUPAC Name (Z)-2-ethylhex-2-enal
Molecular Weight (g/mol) 126.199
PubChem CID 10997035
Formula Weight 126.20
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-2-hexenal (cis- and trans- mixture)
Show More Show Less
EINECSNumber (2)-0513
RTECSNumber MP6300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.