accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6450-57-3

CH3H3COOH3CCH3

CAS RN 6450-57-3

IUPAC Name: 1-(2,4,6-trimethylphenyl)butane-1,3-dione
Synonyms: 1-(2,4,6-trimethylphenyl)butane-1,3-dione
Molecular Weight (g/mol): 204.27
Molecular Formula: C13H16O2
InChi Key: MBZZDUHWYNESIY-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)C1=C(C)C=C(C)C=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results