Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Mesitylene)-1,3-butanedione 96.0+%, TCI America™
SDP

Catalog No. M12725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12725G 5 g
M127225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12725G Supplier TCI America Supplier No. M12725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6450-57-3
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00477603
InChI Key MBZZDUHWYNESIY-UHFFFAOYSA-N
Synonym 1-(2,4,6-Trimethylbenzene)-1,3-butanedione, (2,4,6-Trimethylbenzoyl)acetone
PubChem CID 367102
IUPAC Name 1-(2,4,6-trimethylphenyl)butane-1,3-dione
SMILES CC1=CC(=C(C(=C1)C)C(=O)CC(=O)C)C

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 172°C
Quantity 5 g
Formula Weight 204.27
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Mesitylene)-1,3-butanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.