accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3H3CH3CClBrBr

CAS RN 64531-21-1

IUPAC Name: dibromochloranuide; tetrabutylazanium
Synonyms: dibromochloranuide; tetrabutylammonium ion
Molecular Weight (g/mol): 437.73
Molecular Formula: C16H36Br2ClN
InChi Key: LSCNDUNMPYESRN-UHFFFAOYSA-N
SMILES: Br[Cl-]Br.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results