Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Dibromochloride 96.0+%, TCI America™
SDP

Catalog No. T118925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T118925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T118925G Supplier TCI America Supplier No. T118925G

Chemical Identifiers

CAS 64531-21-1
Molecular Formula C16H36Br2ClN
Molecular Weight (g/mol) 437.729
MDL Number MFCD00060053
InChI Key LSCNDUNMPYESRN-UHFFFAOYSA-N
PubChem CID 23500174
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[Cl-](Br)Br

Specifications

Melting Point 67°C
Quantity 25 g
Formula Weight 437.73
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Dibromochloride
EINECSNumber (2)-0186
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.