accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6459-59-2

IUPAC Name: hydrogen (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid chloride
Synonyms: histidine hydrochloride
Molecular Weight (g/mol): 191.62
Molecular Formula: C6H10ClN3O2
InChi Key: QZNNVYOVQUKYSC-JEDNCBNOSA-N
SMILES: [H+].[Cl-].N[C@@H](CC1=CN=CN1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results