Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Histidine, 99%, MP Biomedicals™

Catalog No. ICN10195105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10195105 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10195105 Supplier MP Biomedicals, Inc Supplier No. 0210195105

missing translation for 'validMainframeMsg'

Specifications

CAS 6459-59-2
Assay Percent Range 99%
Molecular Formula C6H10ClN3O2
MDL Number MFCD00064555
Synonym hydrogen dl-histidine chloride, 2-amino-3-1h-imidazol-5-yl propanoic acid; hydron; chloride, 2-azanyl-3-1h-imidazol-5-yl propanoic acid; hydron; chloride
InChI Key QZNNVYOVQUKYSC-JEDNCBNOSA-N
SMILES [H+].[Cl-].N[C@@H](CC1=CN=CN1)C(O)=O
IUPAC Name hydrogen (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid chloride
Quantity 5 g
Molecular Weight (g/mol) 191.62
PubChem CID 53395185
Formula Weight 209.6
Physical Form Crystalline Powder
Chemical Name or Material DL-Histidine
Show More Show Less
EINECSNumber 229-266-8
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.