accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 646-20-8

IUPAC Name: heptanedinitrile
Synonyms: heptanedinitrile
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: LLEVMYXEJUDBTA-UHFFFAOYSA-N
SMILES: N#CCCCCCC#N
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)