Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pimelonitrile 98.0+%, TCI America™
SDP

Catalog No. P0049500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0049500ML 500 mL
P004925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0049500ML Supplier TCI America Supplier No. P0049500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 646-20-8
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00001980
InChI Key LLEVMYXEJUDBTA-UHFFFAOYSA-N
Synonym 1,5-Dicyanopentane, Heptanedinitrile, Pentamethylene Dicyanide
PubChem CID 12590
IUPAC Name heptanedinitrile
SMILES N#CCCCCCC#N

Specifications

Color Yellow
Boiling Point 175°C
Quantity 500 mL
UN Number 3276
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pimelonitrile
EINECSNumber (2)-4151
RTECSNumber MI9020000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.