accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64603-90-3

IUPAC Name: hydrogen 1,2,3,6-tetrahydropyridine-4-carboxylic acid chloride
Synonyms: isoguvacine
Molecular Weight (g/mol): 163.6
Molecular Formula: C6H10ClNO2
InChi Key: SUWREQRNTXCCBL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC(=O)C1=CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results