Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoguvacine hydrochloride, 99%, Thermo Scientific™

Catalog No. AAJ63243MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63243MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63243MC Supplier Thermo Scientific Chemicals Supplier No. J63243MC

missing translation for 'validMainframeMsg'

Specific GABA agonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Isoguvacine·HCl is an agonist of γ-amino butyric acid receptor. Isoguvacine interactions have many similar biochemical features of GABA synaptic function. Isoguvacine·HCl is a specific GABAA receptor agonist. This compound supp
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64603-90-3
Molecular Formula C6H10ClNO2
Molecular Weight (g/mol) 163.60
MDL Number MFCD01365699
InChI Key SUWREQRNTXCCBL-UHFFFAOYSA-N
Synonym isoguvacine, 1,2,3,6-tetrahydro-pyridine-4-carboxylic acid, 1,2,3,6-tetrahydro-4-pyridinecarboxylic acid, unii-ytf580771y, chembl39071, 4-pyridinecarboxylic acid, 1,2,3,6-tetrahydro, 1,2,3,6-tetrahydropyridinium-4-carboxylate, 1,2,3,6-tetrahydropyridin-1-ium-4-carboxylate, artemisinin sp, spectrum_001974
PubChem CID 3765
ChEBI CHEBI:34799
IUPAC Name 1,2,3,6-tetrahydropyridine-4-carboxylic acid
SMILES [H+].[Cl-].OC(=O)C1=CCNCC1

Specifications

Color White
Quantity 100 mg
Solubility Information Soluble in water
Formula Weight 163.6
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Isoguvacine hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber US5908286
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.