accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBrBrF

CAS RN 64695-78-9

IUPAC Name: 1,2-dibromo-4,5-difluorobenzene
Synonyms: 1,2-dibromo-4,5-difluorobenzene
Molecular Weight (g/mol): 271.89
Molecular Formula: C6H2Br2F2
InChi Key: JTEZQWOKRHOKDG-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C(Br)C=C1F
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (6)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)