Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromo-4,5-difluorobenzene, 99%, Thermo Scientific™

Catalog No. AC249490010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC249490010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC249490010 Supplier Thermo Scientific Chemicals Supplier No. 249490010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64695-78-9
Molecular Formula C6H2Br2F2
Molecular Weight (g/mol) 271.887
MDL Number MFCD00009890
InChI Key JTEZQWOKRHOKDG-UHFFFAOYSA-N
Synonym benzene, 1,2-dibromo-4,5-difluoro, 1,2-dibromo-4,5-difluoro-benzene, 1,2-difluoro-4,5-dibromobenzene, zlchem 536, pubchem3440, acmc-1biw8, 1,2-dibromo-4,5-difluorobenzene, benzene, 1,2-dibromo-4,5-difluoro-;1,2-dibromo-4,5-difl
PubChem CID 94949
IUPAC Name 1,2-dibromo-4,5-difluorobenzene
SMILES C1=C(C(=CC(=C1Br)Br)F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 32°C to 35°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula Br2C6H2F2
Quantity 1 g
Formula Weight 271.88
Percent Purity ≥98.5%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 1, 2-Dibromo-4, 5-difluorobenzene, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.