CAS RN 64742-82-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Petroleum ether, Ultra Resi-analyzed, For ACS analysis, J.T. Baker™
CAS: 64742-82-1 Molecular Formula: C6H14 Synonym: Petrol; Benzin,Benzin