Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum ether, Ultra Resi-analyzed, For ACS analysis, J.T. Baker™

Catalog No. 14650218 Shop All Avantor Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-650-218 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-650-218 Supplier Avantor Supplier No. 926503
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64742-82-1
Molecular Formula C6H14
Synonym Petrol; Benzin, Benzin

Specifications

Melting Point -95°C
Color Colorless
Density 0.77g/mL at 20°C
Quantity 4 L
Physical Form Liquid
Boiling Point 90°C to 100°C
Formula Weight 86.18
Chemical Name or Material Petroleum ether, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.