accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHNNCl

CAS RN 6478-73-5

IUPAC Name: 5,6-dichloro-1H-1,3-benzodiazole
Synonyms: 5,6-dichloro-1H-1,3-benzodiazole
Molecular Weight (g/mol): 187.02
Molecular Formula: C7H4Cl2N2
InChi Key: IPRDZAMUYMOJTA-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C=C2N=CNC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results