CAS RN 648449-76-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PI 3-Kγ Inhibitor II
The PI 3-Kγ Inhibitor II, also referenced under CAS 648449-76-7, controls the biological activity of PI 3-Kγ. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.