accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64960-75-4

IUPAC Name: 2-amino-4-(tert-butoxy)-4-oxobutanoic acid
Synonyms: 2-amino-4-(tert-butoxy)-4-oxobutanoic acid
Molecular Weight (g/mol): 189.21
Molecular Formula: C8H15NO4
InChi Key: MXWMFBYWXMXRPD-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CC(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)