accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65-82-7

IUPAC Name: (2S)-2-acetamido-4-(methylsulfanyl)butanoic acid
Synonyms: N-acetylmethionine
Molecular Weight (g/mol): 191.25
Molecular Formula: C7H13NO3S
InChi Key: XUYPXLNMDZIRQH-LURJTMIESA-N
SMILES: CSCC[C@H](NC(C)=O)C(O)=O
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)