Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-L-methionine, 98+%, Thermo Scientific™

Catalog No. AC226860050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC226860050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC226860050 Supplier Thermo Scientific Chemicals Supplier No. 226860050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-82-7
Molecular Formula C7H13NO3S
Molecular Weight (g/mol) 191.25
MDL Number MFCD00064441
InChI Key XUYPXLNMDZIRQH-LURJTMIESA-N
Synonym n-acetyl-l-methionine, n-acetylmethionine, acetyl-l-methionine, ac-met-oh, l-n-acetyl methionine, acetylmethionine, methionamine, methionine, n-acetyl-, l, thiomedon, l-methionine, n-acetyl
PubChem CID 448580
ChEBI CHEBI:21557
IUPAC Name (2S)-2-acetamido-4-(methylsulfanyl)butanoic acid
SMILES CSCC[C@H](NC(C)=O)C(O)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color White
Ignition Residue 0.1% max.
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Linear Formula CH3SCH2CH2CH(NHCOCH3)CO2H
Quantity 5 g
Merck Index 15, 92
Specific Rotation -19° to -21° (20°C, 589nm) (c=4, H2O)
Formula Weight 191.24
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-methionine, 98+%
Show More Show Less
EINECSNumber 200-617-7
RTECSNumber PD0480000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.