CAS RN 65-85-0
Benzoic acid, 99%
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.123 MDL Number: MFCD00002398 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
Benzoic acid, ACS, 99.5% min
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.123 MDL Number: MFCD00002398 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
Benzoic Acid (Crystalline/Certified ACS), Fisher Chemical
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.123 MDL Number: MFCD00002398 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
Benzoic acid, 99.6%, ACS reagent, Thermo Scientific Chemicals
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.12 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
Benzoic acid, 99%, extra pure
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.12 MDL Number: MFCD00002398 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
Benzoic Acid, Crystal, Reagent, ACS, 99.5%, Spectrum™ Chemical
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.12 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N IUPAC Name: benzoic acid SMILES: OC(=O)C1=CC=CC=C1
Benzoic Acid, FCC, 99.5-100.5%, Spectrum™ Chemical
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.12 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N IUPAC Name: benzoic acid SMILES: OC(=O)C1=CC=CC=C1
Benzoic Acid, ACS Reagent Grade, ≥99.5%, MP Biomedicals™
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.123 MDL Number: MFCD00002398 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
Benzoic Acid Standard, For Elementary Analysis, MilliporeSigma™
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.123 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
Benzoic Acid, Crystal, EP, BP, JP, USP, 99.5-100.5%, Spectrum™ Chemical
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.12 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N IUPAC Name: benzoic acid SMILES: OC(=O)C1=CC=CC=C1
Benzoic acid, 99.5%, for analysis
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.123 MDL Number: MFCD00002398 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
Benzoic Acid Standard, For Quantitative NMR, MilliporeSigma™ Supelco™
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Benzoic Acid, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Benzoic Acid 99.0+%, TCI America™
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.123 MDL Number: MFCD00002398 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N Synonym: benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
Benzoic Acid, SPEX CertiPrep™
CAS: 65-85-0 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.123 InChI Key: WPYMKLBDIGXBTP-UHFFFAOYSA-N PubChem CID: 243 ChEBI: CHEBI:30746 IUPAC Name: benzoic acid SMILES: C1=CC=C(C=C1)C(=O)O
- 1
- 2