Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoic Acid Standard, For Quantitative NMR, MilliporeSigma™ Supelco™

Catalog No. 111018619 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8619 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8619 Supplier MilliporeSigma Supelco Supplier No. 061851G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

Chemical Shift: 7.4 - 8.2 ppm (chemical shifts may slightly vary depending on the experimental conditions)suitable NMR solvents: MeOD, DMSO-d6
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzoic Acid
CAS 65-85-0
Product Line TraceCERT
Vapor Pressure 10 mmHg (132°C)
Vapor Density 4.21 (vs. air)
Physical Form Crystalline
Linear Formula C6H5COOH
Technique Gas Chromatography
MDL Number MFCD00002398
Certifications/Compliance ISO/IEC 17025
Melting Point 121°C to 125°C
Boiling Point 249°C
Industry Type Food and Beverage
Molecular Weight (g/mol) 122.12
Quantity 1 g
Formula Weight 122.12
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.