CAS RN 65005-57-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ NG,NG-Dimethyl-L-arginine, Dihydrochloride
A cell-permeable, reversible inhibitor of nitric oxide synthase in vitro (IC50 = 2-3 μM) and in vivo.