Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NG,NG-Dimethyl-L-arginine, Dihydrochloride

Catalog No. 31120325MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-120-325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-120-325MG Supplier MilliporeSigma™ Supplier No. 31120325MG
Add to Cart
Add to Cart

A cell-permeable, reversible inhibitor of nitric oxide synthase in vitro (IC50 = 2-3 μM) and in vivo.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins NOS in vitro
CAS 65005-57-4
Molecular Formula C8H18N4O2·2HCl
Formula Weight 275.2g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Amorphous Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.