accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CS

CAS RN 65016-61-7

IUPAC Name: 3-heptylthiophene
Synonyms: 3-heptylthiophene
Molecular Weight (g/mol): 182.33
Molecular Formula: C11H18S
InChi Key: IUUMHORDQCAXQU-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CSC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results